missing translation for 'onlineSavingsMsg'
Learn More

Inconel™ 600 gauze, 40 mesh woven from 0.25mm (0.01 in.) dia. wire, Thermo Scientific Chemicals

25.43 € - 132.66 €

Specifications

Chemical Name or Material Inconel™ 600 gauze
Name Note 40 mesh woven from 0.25mm (0.01 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr:Fe; 77:15.5:7.5 wt%
MDL Number MFCD01741927
Solubility Information Insoluble in water.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15407305
View Documents
Thermo Scientific Alfa Aesar
046745.FL
75 x 75 mm
25.43 €
Each
Please to purchase this item. Need a web account? Register with us today!
15417305
View Documents
Thermo Scientific Alfa Aesar
046745.GT
150 x 150 mm
42.01 €
Each
Please to purchase this item. Need a web account? Register with us today!
15427305
View Documents
Thermo Scientific Alfa Aesar
046745.HB
300 x 300 mm
132.66 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications include heaters, stills, bubble towers and condensers for processing of fatty acids; evaporator tubes, tube sheets and flaking trays for the manufacture of sodium sulfide; and equipment for handling abietic acid in the manufacture of paper pulp. It is used for retorts, muffles, roller hearths and other furnace components and for heat-treating baskets and trays. The alloy is a standard material of construction for nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Inconel™ 600 gauze
Ni:Cr:Fe; 77:15.5:7.5 wt%
Insoluble in water.
Wire Cloth
40 mesh woven from 0.25mm (0.01 in.) dia. wire
MFCD01741927
40 Mesh
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350-H372

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.