Learn More
OHAUS™ Incubating Shaker
Save Incubator Space with Benchtop Incubating and Incubating Refrigerating Shakers
Brand:  OHAUS™ 30573775
8566.27 EUR valid until 2024-12-31
Use promo code "21970" to get your promotional price.
 View more versions of this product
Product Code. 17268380
  • 9360.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The exclusive Opti-Flow Forced Ventilation System utilizes twin induction or a single large fan
  • Air is deflected around & not directly on samples, creating a circular flow of air ensuring uniformity
  • For repeatable results, the exclusive Accu-Drive shaking system ensures speed accuracy
  • The Triple Eccentric Drive provides consistent shaking motion throughout the entire speed range
  • The chambers for the ISHD23HDG and ISHD23CDG units are large enough to hold 2 x 6 L Erlenmeyer flasks with optional clamps and platform.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
22.7 kg at 125 rpm, 4.53 kg at 500 rpm
230 V
800 W
TUV
Aluminum, Cycoloy
23.5 in.
26.6 in.
224.07 lb.
ISHD23HDG
Cell Cultures, Solubility Studies, Bacterial Suspensions, Extraction Procedures
1 sec. to 160 hr.
15 to 500 rpm
15°C to 32°C
50 lb. at 125 rpm, 10 lb. at 500 rpm
50/60 Hz
815 x 675 x 596 mm
32.1 in.
103.4 kg
101.6 kg
Independent LED displays for temperature, speed and time
80%, Non-condensing