Learn More
Unity Lab Services™ Incubator Requalification Service with Temperature Mapping
Requalification Services including 1 Temperature Mapping for Incubator (documentation included)
Brand:  Unity Lab Services™ RQTMInc
Product Code. 30178683
  • 3175.00 € / 1 unit
Explore more special offers
Description
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests include door ajar, power failure, and temperature alerts.
  • Temperature mapping (TM): A mapping study that establishes the temperature distribution within the chamber being mapped and locates hot and cold spots, which provides a view of the equipments performance.
  • Temperature measurements will be performed in the air or with simulated products loaded in a chamber, with a data collection interval configurable to customer needs. The collected data provide essential information, helping ensure all material is correctly stored within its labeled temperature range.
  • TM includes one open door temperature recovery test, one power failure test, minimum and maximum values along with averages, a temperature graph over time, and detailed reports that include raw probe data.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Incubator
Requalification Service