Learn More
Mettler Toledo™ Indicating Half-Cell Ion-Selective Electrodes
For quick and easy determination of concentration of a variety of dissolved ions. Mettler Toledo Indicating Half-Cell Ion-Selective Electrodes have sturdy polyoxymethylene (POM) bodies.
Brand:  Mettler Toledo™ 51340900
 View more versions of this product
Product Code. 10312671
  • 1170.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Requires : All ion-selective half cells require InLab™ 302 reference cell
Description
Description
  • Consist of a universal shaft and interchangeable ion-specific membrane module, allowing you to measure ion of your choice
  • All ion-selective half cells require InLab™ 302 reference cell (see 01-920-101, available separately), and appropriate S7 connection cable
Order Info
All are 4.75 in. long (120mm) and 0.5 in. (12mm) in diameter. For other ion-selective electrodes and membrane kits not listed here, contact your Fisher Scientific™ Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Determination of Ammonium concentration of samples such as boiler feed water, food and fertilizer
S7
0.5 in.
4.75 in.
2 to 9
1 to 1E-05mol/L
Polymer Matrix
0°C to 50°C
12 mm
120 mm
0°F to 122°F