Learn More
Mettler Toledo™ Indicating Half-Cell Ion-Selective Electrodes
For quick and easy determination of concentration of a variety of dissolved ions. Mettler Toledo Indicating Half-Cell Ion-Selective Electrodes have sturdy polyoxymethylene (POM) bodies.
Brand:  Mettler Toledo™ 51340500
957.66 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 10414641
  • 1170.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Requires : All ion-selective half cells require InLab™ 302 reference cell.
Description
Description
  • Consist of a universal shaft and interchangeable ion-specific membrane module, allowing you to measure ion of your choice
  • All ion-selective half cells require InLab™ 302 reference cell (see 01-920-101, available separately), and appropriate S7 connection cable
Order Info
All are 4.75 in. long (120mm) and 0.5 in. (12mm) in diameter. For other ion-selective electrodes and membrane kits not listed here, contact your Fisher Scientific™ Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Determination of Fluoride concentration in samples such as drinking water, milk, plants and toothpaste
S7
0.5 in.
4.75 in.
4 to 10
1 to 1E-05mol/L
Polymer Membrane
0°C to 50°C
12 mm
120 mm
0°F to 176°F