Learn More
Thermo Scientific Chemicals Indigo Carmine, High Purity Biological Stain
39.70 € - 378.89 €
Chemical Identifiers
CAS | 860-22-0 |
---|---|
Molecular Formula | C16H8N2Na2O8S2 |
Molecular Weight (g/mol) | 466.35 |
MDL Number | MFCD00005723 |
InChI Key | KHLVKKOJDHCJMG-QDBORUFSSA-L |
Synonym | Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt |
PubChem CID | 5284351 |
IUPAC Name | disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate |
SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10724471
|
Thermo Scientific Chemicals
157440250 |
25 g | Glass bottle |
39.70 €
25g |
In Stock
Log in to see stock. |
||||
10215680
|
Thermo Scientific Chemicals
157441000 |
100 g | Glass bottle |
100.50 €
100g |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
10734471
|
Thermo Scientific Chemicals
157445000 |
500 g | Glass bottle |
378.89 €
500g |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
860-22-0 | |
466.35 | |
KHLVKKOJDHCJMG-QDBORUFSSA-L | |
5284351 | |
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
C16H8N2Na2O8S2 | |
MFCD00005723 | |
Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt | |
disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate |
Specifications
860-22-0 | |
C16H8N2Na2O8S2 | |
25, 304 | |
Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt | |
KHLVKKOJDHCJMG-QDBORUFSSA-L | |
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O | |
466.35 | |
466.34 | |
Blue | |
Indigo Carmine |
85% min. | |
MFCD00005723 | |
15, 4982 | |
Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents | |
Authentic | |
disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate | |
5284351 | |
Analysis | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 212-728-8
missing translation for 'rtecsNumber' : DU3000000
missing translation for 'tsca' : TSCA
- Indigo Carmine
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.