missing translation for 'onlineSavingsMsg'
Learn More

Indium, 99.999%, Powder, Thermo Scientific Chemicals

Product Code. 10398710
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Spezifikation

Color Gray
Melting Point 156.0°C
Boiling Point 2000.0°C
Density 7.3000g/mL
Assay Percent Range 100.00%
Linear Formula In
Quantity 5 g
Fieser 06,293; 14,181; 16,181; 17,152
Formula Weight 114.82
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Form Powder
Chemical Name or Material Indium
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Indium, powder
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Flammable solid Category 2
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'