missing translation for 'onlineSavingsMsg'
Learn More

Indium Arsenide, 99.9999% (metals basis), Thermo Scientific Chemicals

139.70 € - 513.56 €

Chemical Identifiers

CAS 1303-11-3
Molecular Formula AsIn
Molecular Weight (g/mol) 189.74
MDL Number MFCD00016144
InChI Key RPQDHPTXJYYUPQ-UHFFFAOYSA-N
Synonym indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g
PubChem CID 91500
IUPAC Name indiganylidynearsane
SMILES [As]#[In]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11372018
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012932.04
2 g
139.70 €
2g
Please to purchase this item. Need a web account? Register with us today!
11392018
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012932.09
10 g
513.56 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indium arsenide is used for construction of infrared detectors, for the wavelength range of 1-3μm. It is widely used as terahertz radiation source as it is a strong photo-Dember emitter. Indium arsenide is also used for making of diode laser.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1303-11-3
189.74
RPQDHPTXJYYUPQ-UHFFFAOYSA-N
91500
[As]#[In]
AsIn
MFCD00016144
indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g
indiganylidynearsane

Specifications

936°C
Gray
99.9999% (Metal basis)
MFCD00016144
Insoluble in water.
[As]#[In]
189.74
189.74
Gray polycrystalline pieces
1303-11-3
Pieces
AsIn
indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g
RPQDHPTXJYYUPQ-UHFFFAOYSA-N
indiganylidynearsane
91500
Odorless
Indium arsenide, Hardly attacked by mineral acids
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 215-115-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only