missing translation for 'onlineSavingsMsg'
Learn More

Indium foil, 0.5mm (0.02in) thick, Puratronic™, 99.9975% (metals basis)

Product Code. 11351179
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
50 x 100 mm
100 x 200 mm
Unit Size:
25mm
Each
This item is not returnable. View return policy

Product Code. 11351179

Brand: Thermo Scientific Alfa Aesar 011384.FY

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Quantity 50 x 100 mm
Odor Odorless
Weight ≈2.3g/25x25mm
Form Foil
Percent Purity 99.9975%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Indium foil, 0.5mm (0.02 in.) thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.