missing translation for 'onlineSavingsMsg'
Learn More

Indium(I) bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11330108
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14280-53-6
Molecular Formula InBr
MDL Number MFCD00064630
Synonym indium i bromide, indium 1+ ;bromide, acmc-1bzdu, indium bromide inbr 6ci,7ci,8ci,9ci

Specifications

Boiling Point 662°C
Melting Point 220°C
Physical Form Polycrystalline Lump
Quantity 25 g
UN Number UN3260
Formula Weight 194.73
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Sensitivity Moisture sensitive
Density 4.96 g/mL
Assay (metals basis)
Chemical Name or Material Indium (I) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'