missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) bromide, 99.99%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10350614
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-09-3
Molecular Formula Br3In
Molecular Weight (g/mol) 354.52
MDL Number MFCD00011057

Specifications

Boiling Point 656.0°C
Melting Point 220.0°C
Color White
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99.99%
Linear Formula InBr3
Merck Index 15, 4997
Formula Weight 354.52
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Indium(III) bromide
Show More Show Less
compliance-icons
Product Identifier
  • Indium(III) bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'