Learn More
Indium(III) bromide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Indium(III) bromide is used in organic synthesis as a water tolerant Lewis acid.It efficiently catalyzes the three-component coupling of β-keto esters, aldehydes and urea (or thiourea) to afford the corresponding dihydropyrimidinones. It is used as a catalyst to produce dithioacetals when unactivated alkynes react with thiols and fields such as optics and microelectronics that utilize semiconductor technology have wide uses for indium in high-performing solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∼436°C |
Color | White to Yellow |
Physical Form | Crystalline Powder and/or Chunks |
Quantity | 10 g |
Linear Formula | InBr3 |
Solubility Information | Soluble in water. |
Formula Weight | 354.55 |
Percent Purity | 99.99% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
W25E108 | Certificate of Analysis | 25/11/2024 | 040126.18,040126.09 |
U07K047 | Certificate of Analysis | 22/08/2024 | 040126.18,040126.09 |
M02K034 | Certificate of Analysis | 10/01/2024 | 040126.18,040126.09 |
T02F012 | Certificate of Analysis | 03/01/2024 | 040126.18,040126.09 |
U11J030 | Certificate of Analysis | 24/10/2023 | 040126.18,040126.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.