missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride, White to off-white powder, 99.999% (Metals basis), Thermo Scientific Chemicals

Product Code. 11428490
Click to view available options
Quantity:
5 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Indium(III) chloride is used as catalyst for aqueous organic reactions including C-C bond formation, aldol reactions, and reductions and for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-82-8
Molecular Formula InCl3
MDL Number MFCD00011058

Specifications

Boiling Point 800°C
Melting Point 586°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Reacts with water.
Formula Weight 221.18
Sensitivity Hygroscopic
Density 3.46
Chemical Name or Material Indium(III) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'