missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) hydroxide, 99.8% (metals basis), Thermo Scientific Chemicals

60.20 € - 147.74 €

Chemical Identifiers

CAS 20661-21-6
MDL Number MFCD00011056
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329237
View Documents
Thermo Scientific Alfa Aesar
040100.09
10 g
60.20 €
10g
Please to purchase this item. Need a web account? Register with us today!
11319237
View Documents
Thermo Scientific Alfa Aesar
040100.18
50 g
147.74 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indium(III) hydroxide is used as a precursor to indium(III)oxide. It is also applied in batteries, glasses, ceramics, chemical reagents and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20661-21-6
MFCD00011056

Specifications

150°C (in -H2O)
Powder
Insoluble in water.
99.8%
Indium(III) hydroxide
20661-21-6
MFCD00011056
165.84
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 243-947-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only