missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) iodide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

102.51 € - 904.50 €

Chemical Identifiers

CAS 13510-35-5
Molecular Formula I3In
Molecular Weight (g/mol) 495.531
MDL Number MFCD00016151
InChI Key RMUKCGUDVKEQPL-UHFFFAOYSA-K
Synonym indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83539
IUPAC Name triiodoindigane
SMILES [In](I)(I)I
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371918
View Documents
Thermo Scientific Alfa Aesar
011854.04
2 g
102.51 €
2g
Please to purchase this item. Need a web account? Register with us today!
11391918
View Documents
Thermo Scientific Alfa Aesar
011854.09
10 g
348.74 €
10g
Please to purchase this item. Need a web account? Register with us today!
11381918
View Documents
Thermo Scientific Alfa Aesar
011854.18
50 g
904.50 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indium(III) iodide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13510-35-5
495.531
RMUKCGUDVKEQPL-UHFFFAOYSA-K
83539
[In](I)(I)I
I3In
MFCD00016151
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
triiodoindigane

Specifications

210°C
Red to Yellow
99.994% (Metals basis)
MFCD00016151
Soluble in water.
[In](I)(I)I
495.531
495.53
4.69
13510-35-5
Crystalline
I3In
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
RMUKCGUDVKEQPL-UHFFFAOYSA-K
triiodoindigane
83539
Hygroscopic
Indium(III) iodide
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335-H360

missing translation for 'einecsNumber' : 236-839-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.