missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) n-octoxide, 98%, Thermo Scientific™

Product Code. 15477787
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106461-03-4
MDL Number MFCD26967910
Synonym indium 3+ ion octanol

Specifications

Melting Point ∼150°C (decomposition)
Color Yellow
Physical Form Solid
Quantity 5 g
UN Number UN1325
Formula Weight 502.48
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material Indium(III) n-octoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'