Learn More
Indium(III) nitrate hydrate, In 29% min., Thermo Scientific Chemicals
148.74 € - 371.85 €
Chemical Identifiers
CAS | 207398-97-8 |
---|---|
Molecular Formula | InN3O9 |
Molecular Weight (g/mol) | 300.83 |
MDL Number | MFCD00149742 |
InChI Key | LKRFCKCBYVZXTC-UHFFFAOYSA-N |
Synonym | indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2oShow More |
PubChem CID | 3084148 |
IUPAC Name | indium(3+) trinitrate |
SMILES | [In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11473787
|
Thermo Scientific Alfa Aesar
A18252.14 |
25 g |
148.74 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11483787
|
Thermo Scientific Alfa Aesar
A18252.22 |
100 g | N/A | N/A | N/A | |||||
Description
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIndium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.
Solubility
Soluble in water, methanol and ethanol.
Notes
It is stable at room temperature. Hygroscopic. Incompatible with strong reducing agents and strong acids.
Chemical Identifiers
207398-97-8 | |
300.83 | |
LKRFCKCBYVZXTC-UHFFFAOYSA-N | |
3084148 | |
[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
InN3O9 | |
MFCD00149742 | |
indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2oShow More | |
indium(3+) trinitrate |
Specifications
100°C (in 2H2O) | |
InN3O9 | |
UN1477 | |
Soluble in water,methanol and ethanol. | |
[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
300.83 | |
300.83 (Anhydrous) | |
In ≥29% |
207398-97-8 | |
MFCD00149742 | |
indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2o | |
LKRFCKCBYVZXTC-UHFFFAOYSA-N | |
indium(3+) trinitrate | |
3084148 | |
Hygroscopic | |
Indium(III) nitrate hydrate |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 237-393-5
missing translation for 'tsca' : Yes
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.