missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 11370979
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Melting Point 850°C (volat.)
Physical Form Powder
Quantity 50 g
Merck Index 14,4952
Formula Weight 277.64
Percent Purity 99.997%
Grade Puratronic™
Odor Odorless
Density 7.18 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'