Learn More
Indium(III) phosphide, 99.9999% (metals basis), Thermo Scientific Chemicals
196.98 € - 884.40 €
Chemical Identifiers
CAS | 22398-80-7 |
---|---|
Molecular Formula | InP |
Molecular Weight (g/mol) | 145.792 |
MDL Number | MFCD00016153 |
InChI Key | GPXJNWSHGFTCBW-UHFFFAOYSA-N |
Synonym | indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 |
PubChem CID | 31170 |
IUPAC Name | indiganylidynephosphane |
SMILES | P#[In] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331489
|
Thermo Scientific Alfa Aesar
013974.03 |
1 g |
196.98 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321489
|
Thermo Scientific Alfa Aesar
013974.06 |
5 g |
884.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Indium(III) phosphide is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct band gap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22398-80-7 | |
145.792 | |
GPXJNWSHGFTCBW-UHFFFAOYSA-N | |
31170 | |
P#[In] |
InP | |
MFCD00016153 | |
indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 | |
indiganylidynephosphane |
Specifications
1070°C | |
Pieces | |
MFCD00016153 | |
indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 | |
GPXJNWSHGFTCBW-UHFFFAOYSA-N | |
indiganylidynephosphane | |
31170 | |
99.9999% | |
(metals basis) |
22398-80-7 | |
InP | |
14,4953 | |
Insoluble in water. | |
P#[In] | |
145.792 | |
145.79 | |
4.81 g/mL | |
Indium(III) phosphide |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P260-P264b-P270-P281-P308+P313-P501c
H350-H361f-H372
missing translation for 'einecsNumber' : 244-959-5
missing translation for 'rtecsNumber' : NL1800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only