missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis), Thermo Scientific Chemicals

69.04 € - 1128.00 €

Chemical Identifiers

CAS 22398-80-7
Molecular Formula InP
Molecular Weight (g/mol) 145.792
MDL Number MFCD00016153
InChI Key GPXJNWSHGFTCBW-UHFFFAOYSA-N
Synonym indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875
PubChem CID 31170
IUPAC Name indiganylidynephosphane
SMILES P#[In]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11472907
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
A15509.03
1 g
69.04 €
1g
Please to purchase this item. Need a web account? Register with us today!
11482907
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
A15509.06
5 g
258.29 €
5g
Please to purchase this item. Need a web account? Register with us today!
11492907
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
A15509.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications

Chemical Identifiers

22398-80-7
145.792
GPXJNWSHGFTCBW-UHFFFAOYSA-N
31170
P#[In]
InP
MFCD00016153
indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875
indiganylidynephosphane

Specifications

1070°C
Polycrystalline Lump
MFCD00016153
indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875
GPXJNWSHGFTCBW-UHFFFAOYSA-N
indiganylidynephosphane
31170
99.99%
(metals basis)
22398-80-7
InP
14,4953
Insoluble in water.
P#[In]
145.792
145.79
4.81
Indium(III) phosphide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P260-P264b-P270-P281-P308+P313-P501c

H350-H361f-H372

missing translation for 'einecsNumber' : 244-959-5

missing translation for 'rtecsNumber' : NL1800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only