Learn More
Indium(III) sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
156.78 € - 518.58 €
Chemical Identifiers
CAS | 13464-82-9 |
---|---|
Molecular Formula | In2O12S3 |
Molecular Weight (g/mol) | 517.80 |
MDL Number | MFCD00011059 |
InChI Key | XGCKLPDYTQRDTR-UHFFFAOYSA-H |
Synonym | indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german |
PubChem CID | 26044 |
SMILES | [In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349237
|
Thermo Scientific Alfa Aesar
040102.14 |
25 g |
156.78 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339237
|
Thermo Scientific Alfa Aesar
040102.22 |
100 g |
518.58 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Indium(III) sulfate, anhydrous is used in the preparation of indium or indium containing substances like copper indium selenide. It is employed to electroplate indium metal, as a hardening agent in gold electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13464-82-9 | |
517.80 | |
XGCKLPDYTQRDTR-UHFFFAOYSA-H | |
26044 |
In2O12S3 | |
MFCD00011059 | |
indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german | |
[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Specifications
13464-82-9 | |
Solid | |
MFCD00011059 | |
Soluble in water. | |
[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
517.80 | |
517.83 | |
Odorless | |
3.438 g/mL | |
Indium(III) sulfate, Anhydrous |
Gray | |
In2O12S3 | |
indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german | |
XGCKLPDYTQRDTR-UHFFFAOYSA-H | |
indium(3+);trisulfate | |
26044 | |
99.99% | |
Hygroscopic | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 236-689-1
missing translation for 'rtecsNumber' : NL1925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.