missing translation for 'onlineSavingsMsg'
Learn More
Indium(III) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals Product Code.: 11386239

Indium(III) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11386239
50 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11386239

Brand: Thermo Scientific Alfa Aesar 044836.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indium(III) sulfide emerged as a promising low-hazard buffer material and is used to improve the properties of solar cells and also replaces the hazardous CdS. It is a mid band gap semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-24-9
Molecular Formula In2S3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011061
InChI Key SIXIBASSFIFHDK-UHFFFAOYSA-N
IUPAC Name diindium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[In+3].[In+3]

Specifications

Melting Point 1050°C
Color Red to Orange
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water and organic solvents.
Formula Weight 325.83
Percent Purity 99.995%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Indium (III) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN03L043 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog Number044836.09,044836.18,044836.04
Lot NumberZ24K054 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number044836.09,044836.18
Lot NumberZ01J015 Certificate TypeCertificate of Analysis Date17/01/2024 Catalog Number044836.09,044836.18
Lot NumberZ07I031 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number044836.09,044836.18
Lot NumberT12I019 Certificate TypeCertificate of Analysis Date21/07/2022 Catalog Number044836.09,044836.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium(III) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato