missing translation for 'onlineSavingsMsg'
Learn More

Indium ingot, 99.999% (metals basis), Thermo Scientific Chemicals

140.70 € - 434.16 €

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367028
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014720.09
10 g
140.70 €
10g
Please to purchase this item. Need a web account? Register with us today!
11377028
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014720.18
50 g
434.16 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-74-6
114.82
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]

Specifications

White
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
[In]
114.82
CHEBI:30430
Odorless
(metals basis)
7440-74-6
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
5359967
99.999%
Ingot
Indium ingot
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-180-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only