missing translation for 'onlineSavingsMsg'
Learn More
Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals Product Code.: 11349877

Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals

Product Code. 11349877
2 g
Click to view available options
Quantity:
2 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 11349877

Brand: Thermo Scientific Alfa Aesar 042229.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Preparation of transparent electrical conducting electrodes

It is used in the preparation of transparent electrical conducting electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Physical Form Viscous Liquid
Quantity 2 g
UN Number UN1993
Flash Point 13°C (55°F)
Solubility Information Soluble in hexane and toluene; Insoluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈20 wt%
Chemical Name or Material Indium oxide, Tin doped, polymeric precursor

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.