missing translation for 'onlineSavingsMsg'
Learn More

Indium spheres, 1mm (0.04in) dia, 99.99% (metals basis)

Product Code. 11373908
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 11373908

Brand: Thermo Scientific Alfa Aesar 040339.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tolerance: ±0.1mm | CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver-White
Appearance Silvery-white spheres
Quantity 50 g
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Form Sphere
Chemical Name or Material Indium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.