missing translation for 'onlineSavingsMsg'
Learn More

Indium Standard, 1000μg/mL in 2-5% HNO3 (Assurance Grade), SPEX CertiPrep™

Product Code. 10496932
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
In1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-74-6, 7697-37-2, 7732-18-5
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Packaging Triple-leached LDPE bottle
CAS Min % 0.1, 2.0, 97.9
Quantity 125 mL
CAS Max % 0.1, 2.0, 97.9
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration 
 1000μg/mL  2-5% HNO3
Solvent or Matrix 2% HNO3
Chemical Name or Material Indium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'