missing translation for 'onlineSavingsMsg'
Learn More

Indium Tin eutectic ingot, 99.95% (metals basis), Thermo Scientific Chemicals

84.52 € - 368.84 €

Specifications

Chemical Name or Material Indium Tin eutectic ingot
Melting Point 117°C
Avg. Mol. Wt. or Mol. Wt. Range In:Sn; 52:48 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD00144477
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363417
View Documents Promotion Details
Thermo Scientific Alfa Aesar
018159.09
10 g
84.52 €
10g
Please to purchase this item. Need a web account? Register with us today!
11460490
View Documents Promotion Details
Thermo Scientific Alfa Aesar
018159.50
5x 10 g
368.84 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Indium Tin eutectic ingot
In:Sn; 52:48 wt%
MFCD00144477
Ingot
117°C
(metals basis)
99.99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only