missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis)
Click to view available options
Quantity:
5 g
25 g
100 g
Tamaño de la unidad:
100g
25g
5g
Descripción
Indium tin oxide is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Physical Form | Pieces |
| Quantity | 5 g |
| Solubility Information | Insoluble in water. |
| Percent Purity | 99.99% |
| Assay | (metals basis) |
| Chemical Name or Material | Indium tin oxide |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido