missing translation for 'onlineSavingsMsg'
Learn More

Indium wire, 2.0mm (0.08in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Form: Approx.22.95g/m

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359368
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043265.BU
25 cm N/A
N/A
11379368
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043265.G1
1 m N/A
N/A
11369368
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043265.G5
5 m N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-74-6
114.82
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]

Specifications

Silver-White
Silvery-white wire
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
[In]
114.82
CHEBI:30430
Wire
7440-74-6
99.99% (Metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
5359967
Odorless
Indium
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-180-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only