missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium wire, 2.0mm (0.08in) dia, Puratronic™, 99.998+% (metals basis), Thermo Scientific Chemicals
117.00 € - 1501.00 €
Chemical Identifiers
CAS | 7440-74-6 |
---|---|
Molecular Formula | In |
Molecular Weight (g/mol) | 114.82 |
MDL Number | MFCD00134048 |
InChI Key | APFVFJFRJDLVQX-UHFFFAOYSA-N |
Synonym | indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic |
PubChem CID | 5359967 |
ChEBI | CHEBI:30430 |
IUPAC Name | indium |
SMILES | [In] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347107
|
Thermo Scientific Alfa Aesar
011461.BU |
25 cm |
117.00 €
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357107
|
Thermo Scientific Alfa Aesar
011461.G1 |
1 m |
356.00 €
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367107
|
Thermo Scientific Alfa Aesar
011461.G5 |
5 m | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-74-6 | |
114.82 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
5359967 | |
indium |
In | |
MFCD00134048 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
CHEBI:30430 | |
[In] |
Specifications
7440-74-6 | |
MFCD00134048 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
indium | |
5359967 | |
≥99.998% | |
Odorless | |
Wire | |
Indium wire, 2.0mm (0.08 in.) dia. |
In | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
[In] | |
114.82 | |
CHEBI:30430 | |
Puratronic™ | |
≈22.95g/m | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 231-180-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only