missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads, with Pictograms - Flammable/Non Flammable Liquids
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand: Brady™ N008073
This item is not returnable.
View return policy
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC | |
Individual linerless Pipe Markers on a Roll with die-cut arrowheads, with pictograms - Flammable/Non Flammable Liquids/Oils - Diesel | |
Silicone | |
Warning | |
Polyester | |
Diesel | |
English | |
30 Pk. | |
-40°C to +140°C | |
0.119 mm | |
33 m x 450 mm x 52 mm | |
33 m | |
450 mm | |
52 mm |
DIESEL 450X52RL-T1-LL-P19 | |
Permanent Acrylic, Pressure-sensitive | |
White on Dark Brown | |
Roll | |
Pipe Marker | |
Pipe and valve marking | |
30 Markers/Roll | |
-40°F to +284°F | |
0.0047 in. | |
33 m x 450 mm | |
36.09 yd. | |
17.72 in. | |
2.05 in. | |
22.5 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction