Learn More
Brady™ Individual Linerless Pipe Markers on a Roll with Die-cut Arrowheads - Water (A-H)
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand:  Brady™ N008840
 View more versions of this product
Product Code. 15349335
  • 176.00 € / 1 roll
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC
Individual linerless Pipe Markers on a Roll with die-cut arrowheads, without pictograms - Water - Hot water return
Silicone
Roll
Pipe Marker
Pipe and valve marking
30 Markers/Roll
-40°F to +284°F
0.0047 in.
33 m x 450 mm
33 m
450 mm
52 mm
HOT WATER RETURN 450X52RL-T1-LL-P03
Permanent Acrylic, Permanent, Pressure-sensitive
White on Green
Polyester
Hot water return
English
30 Pk.
-40°C to +140°C
0.119 mm
36.09 yd.
17.72 in.
2.05 in.
22.5 mm