missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, with Pictograms, Gas

European Standard Pipemarkers reinforce safety and aid maintenance.

16.50 €

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color Black on Ochre
Finish Gloss
Format Card
View More Specs

Products 18
Product Code Brand Display Text Price Quantity & Availability  
Product Code Brand Display Text Price Quantity & Availability  
15359045
View Documents
Brady™
N007373
Bio gas
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15379025
View Documents
Brady™
N007346
Argon
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15329055
View Documents
Brady™
N007435
CO2
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15359095
View Documents
Brady™
N007531
Carbon dioxide
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15309065
View Documents
Brady™
N007444
Condensate
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15359105
View Documents
Brady™
N007551
Cooling gas
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15369065
View Documents
Brady™
N007466
Exhaust
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15349075
View Documents
Brady™
N007486
Freon
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15309085
View Documents
Brady™
N007495
Freon 11
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15399125
View Documents
Brady™
N007660
Helium
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15309095
View Documents
Brady™
N007513
Gas
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15329085
View Documents
Brady™
N007504
Freon 22
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15389105
View Documents
Brady™
N007571
Inert gas
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15369125
View Documents
Brady™
N007627
LPG
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15319115
View Documents
Brady™
N007580
Mixed gas
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15359115
View Documents
Brady™
N007589
Natural gas
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15389035
View Documents
Brady™
N007355
Nitrogen
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
15319125
View Documents
Brady™
N007618
Recycled gas
16.50 €
Pack of 3
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
Description

Description

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Black on Ochre
Card
Pipe Marker
3
3 Markers/Card
-40°F to +230°F
0.0039 in.
37 x 335 mm
355 mm
37 mm
Permanent Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
Pipe and valve marking
English
3 Pk.
-40°C to +110°C
0.099 mm
13.98 in.
1.46 in.
16.5 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.