Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms - Acids and Alkalis
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand:  Brady™ N007114
Product Code. 15389015
  • 16.50 € / Pack of 3
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
Specifications
Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Acids and Alkalis - Alkali
White on Purple
Card
Pipe Marker
Pipe and valve marking
English
3 Pk.
-40°C to +110°C
0.099 mm
355 mm
37 mm
Permanent Acrylic
Gloss
Polyester
Alkali
3
3 Markers/Card
-40°F to +230°F
0.0039 in.
13.98 in.
1.46 in.
16.5 mm