missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Individual Pipe Markers On A Card with Die-cut Arrowheads, without Pictograms -- Fire Fighting
European Standard Pipemarkers reinforce safety and aid maintenance.
16.50 €
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
---|---|
Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
Color | White on Red |
Finish | Gloss |
Format | Card |
Product Code | Brand | Display Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Price | Quantity & Availability | |||||
15369245
|
Brady™
N008530 |
Fire protection Water |
16.50 €
Pack of 3 |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
15399255
|
Brady™
N008548 |
Foam |
16.50 €
Pack of 3 |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
15359265
|
Brady™
N008584 |
Sprinkler |
16.50 €
Pack of 3 |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
15339255
|
Brady™
N008539 |
Sprinkler Water |
16.50 €
Pack of 3 |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 | |
White on Red | |
Card | |
Pipe Marker | |
3 | |
3 Markers/Card | |
-40°F to +230°F | |
0.0039 in. | |
37 x 335 mm | |
355 mm | |
37 mm |
Permanent Acrylic, Permanent, Pressure-sensitive | |
Gloss | |
Polyester | |
Pipe and valve marking | |
English | |
3 Pk. | |
-40°C to +110°C | |
0.099 mm | |
13.98 in. | |
1.46 in. | |
16.5 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title