Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand:  Fisherbrand™ FB67773
 View more versions of this product
Product Code. 15525059
  • 85.00 € / 1 unit
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class F1 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
Specifications
F1
100 g
Cylindrical
Stainless Steel
100 g