Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1 with certificate
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand:  Fisherbrand™ FB67801
 View more versions of this product
Product Code. 15505089
  • 395.00 € / 1 unit
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class F1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications
Specifications
F1
2 kg
Cylindrical
Stainless Steel
2 kg