Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
33.00 € - 722.00 €
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
15565149
|
Fisherbrand™
FB67867 |
10 g |
42.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15565169
|
Fisherbrand™
FB67887 |
500 g |
118.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15565189
|
Fisherbrand™
FB67907 |
10 kg |
722.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15585139
|
Fisherbrand™
FB67859 |
2 g |
34.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15585159
|
Fisherbrand™
FB67879 |
100 g |
52.00 €
1 unit |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
15585179
|
Fisherbrand™
FB67899 |
5 kg |
391.00 €
1 unit |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
15545139
|
Fisherbrand™
FB67855 |
1 g |
33.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15545159
|
Fisherbrand™
FB67875 |
50 g |
47.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15545179
|
Fisherbrand™
FB67895 |
2 kg |
228.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
15525169
|
Fisherbrand™
FB67883 |
200 g |
66.00 €
1 unit |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
15525149
|
Fisherbrand™
FB67863 |
5 g |
37.00 €
1 unit |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
15505179
|
Fisherbrand™
FB67891 |
1 kg |
153.00 €
1 unit |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
15505159
|
Fisherbrand™
FB67871 |
20 g |
44.00 €
1 unit |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
- Class F2 stainless steel calibration weight
- Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
- In compliance with OIML R111
Specifications
F2 | |
Stainless Steel | |
Cylindrical |