Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand:  Fisherbrand™ FB67869
 View more versions of this product
Product Code. 15585149
  • 16.00 € / 1 unit
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class M1 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
Specifications
M1
10 g
Cylindrical
Stainless Steel
10 g