Learn More
Tocris Bioscience™ Indo 1AM
Fluorescent Ca2+ indicator
Brand:  Tocris Bioscience™ 6704/1
Product Code. 18775416
  • 266.00 € / 1mg
Explore more special offers
Description
Description
Indo 1AM is a fluorescent Ca2+ indicator (Kd for Ca2+ = 250 nM). Cell-permeable. Displays ratiometric emission wavelength change upon Ca2+ binding. Excitation/emission λ are 355/475 nm for no Ca2+ and 355/401 nm for high Ca2+. Absorption maxima are 349 and 331 nm for free anion and Ca complex, respectively. Respective emission maxima are 485 and 410 nm. Quantum yield ~0.5. Ideal for flow cytometry.
Specifications
Specifications
2-[4-[Bis[2-[(acetoxy)methoxy]-2-oxoethyl]amino]-3-[2-[2-[bis[2-[(acetyloxy)methoxy]-2-oxoethyl]amino]-5-methylphenoxy]ethoxy]phenyl]-1H-indole-6-carboxylic acid (acetoxy)methyl ester
1 mg
C47H51N3O22
90%
112926-02-0
Calcium Signaling Probes and Ca2+ Indicator Dyes
1009.91