Learn More
Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Description
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.
Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.

Specifications
Specifications
Melting Point | ∼167°C (decomposition) |
Flash Point | 171°C (339°F) |
Quantity | 25 g |
Beilstein | 143358 |
Sensitivity | Light sensitive |
Merck Index | 14,4964 |
Solubility Information | Soluble in ethanol (50mg/ml),methanol,DMSO,and chloroform (sparingly). Insoluble in water. |
Formula Weight | 175.19 |
Percent Purity | ≥98% |
Chemical Name or Material | Indole-3-acetic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10248453 | Certificate of Analysis | 05/04/2024 | A10556.22,A10556.14,A10556.36,A10556.06 |
10237576 | Certificate of Analysis | 12/04/2022 | A10556.22,A10556.14,A10556.36,A10556.06 |
10105606 | Certificate of Analysis | 22/11/2021 | A10556.22,A10556.14,A10556.36,A10556.06 |
J5993A | Certificate of Analysis | 22/11/2021 | A10556.22,A10556.14,A10556.36,A10556.06 |
10217615 | Certificate of Analysis | 22/11/2021 | A10556.22,A10556.14,A10556.36,A10556.06 |
Safety and Handling

- Indole-3-acetic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 1H-Indole-3-acetic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.