Learn More
Indole-3-butyric acid, MedChemExpress
MedChemExpress Indole-3-butyric acid (3-indolebutyric acid; IBA) is a plant growth auxin and a good rooting agent. It can promote herbs and woody ornamental plant rooting and used for improving fruit rate.
41.85 € - 70.20 €
Specifications
Chemical Name or Material | Indole-3-butyric acid |
---|---|
CAS | 133-32-4 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C12H13NO2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16461328
|
Medchem Express
HY-N0186 |
5 g |
41.85 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16458888
|
Medchem Express
HY-N0186 |
10 g |
70.20 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Indole-3-butyric acid (3-indolebutyric acidSpecifications
Indole-3-butyric acid | |
Off-White | |
C12H13NO2 | |
DMSO : ≥ 35 mg/mL (172.21 mM) | |
203.24 | |
99.9% | |
Solid |
133-32-4 | |
Research | |
3-indolebutyric acid | |
O=C(O)CCCC1=CNC2=C1C=CC=C2 | |
203.24 | |
Research | |
Metabolism-sugar/lipid metabolism |
Safety and Handling
H301∣H315∣H319∣H335∣H412
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.