missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Indole-6-carboxaldehyde, 97%

Product Code. 15421318
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1196-70-9
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD02179596
InChI Key VSPBWOAEHQDXRD-UHFFFAOYSA-N
Synonym indole-6-carboxaldehyde, 6-formylindole, 1h-indole-6-carboxaldehyde, indole-6-carbaldehyde, 6-indolecarboxaldehyde, indole-6-carboxalehyde, 6-formyl-1h-indole, indole-6-carboxaldehyde 6-formylindole, 6-formyl-indole, 6-formyl indole
PubChem CID 2773435
IUPAC Name 1H-indole-6-carbaldehyde
SMILES C1=CC(=CC2=C1C=CN2)C=O

Specifications

Melting Point 127°C to 131°C
Color Brown to White
Quantity 1 g
Formula Weight 145.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Indole-6-carboxaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.