missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Indole-6-carboxaldehyde, 97%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Melting Point | 127°C to 131°C |
Color | Brown to White |
Quantity | 250 mg |
Formula Weight | 145.16 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | Indole-6-carboxaldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction