missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-methanol, 97%, Thermo Scientific™

Product Code. 15575896
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1075-26-9
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD02179595
InChI Key WRMZOPANDOHWJU-UHFFFAOYSA-N
Synonym 6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol
PubChem CID 2773459
SMILES OCC1=CC=C2C=CNC2=C1

Specifications

Melting Point 45.0°C to 50.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name (1H-indol-6-yl)methanol
Formula Weight 147.17
Percent Purity 97%
Chemical Name or Material Indole-6-methanol
compliance-icons
Product Identifier
  • Indol-6-methanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.