missing translation for 'onlineSavingsMsg'
Learn More

Indoline, 99%, Thermo Scientific Chemicals

49.85 € - 595.97 €

Chemical Identifiers

CAS 496-15-1
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00005705
InChI Key LPAGFVYQRIESJQ-UHFFFAOYSA-N
Synonym indoline, 2,3-dihydroindole, 1-azaindan, dihydroindole, 1h-indole, 2,3-dihydro, azaindane, benzopyrrolidine, aza-indane, indoline, 19, pubchem7502
PubChem CID 10328
ChEBI CHEBI:43295
IUPAC Name 2,3-dihydro-1H-indole
SMILES C1CNC2=CC=CC=C21
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11424453
View Documents
Thermo Scientific Alfa Aesar
A11000.18
50 g
49.85 €
50g
Please to purchase this item. Need a web account? Register with us today!
11434453
View Documents
Thermo Scientific Alfa Aesar
A11000.30
250 g
189.95 €
250g
Please to purchase this item. Need a web account? Register with us today!
11414453
View Documents
Thermo Scientific Alfa Aesar
A11000.0B
1000 g
595.97 €
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indole is used in perfumery and in preparing tryptophan, one of the 20 amino acids commonly found in animal proteins. It has important application in the industry of plant growth. It is used to prepare indoleacetic acid (auxin) and other plant growth substances which help the development of roots in plant. Indole and its derivatives are widely used in making perfumes, dyes, agrochemicals and medicines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole is used in perfumery and in preparing tryptophan, one of the 20 amino acids commonly found in animal proteins. It has important application in the industry of plant growth. It is used to prepare indoleacetic acid (auxin) and other plant growth substances which help the development of roots in plant. Indole and its derivatives are widely used in making perfumes, dyes, agrochemicals and medicines.

Solubility
Fully misicible in water at 5 g/L (20°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
Specifications

Chemical Identifiers

496-15-1
119.167
LPAGFVYQRIESJQ-UHFFFAOYSA-N
10328
2,3-dihydro-1H-indole
C8H9N
MFCD00005705
indoline, 2,3-dihydroindole, 1-azaindan, dihydroindole, 1h-indole, 2,3-dihydro, azaindane, benzopyrrolidine, aza-indane, indoline, 19, pubchem7502
CHEBI:43295
C1CNC2=CC=CC=C21

Specifications

496-15-1
1.062
92°C (197°F)
C8H9N
MFCD00005705
Light sensitive
Fully misicible in water at 5g/L (20°C).
C1CNC2=CC=CC=C21
119.167
CHEBI:43295
99%
-21°C
220°C to 221°C
Odorless
1.593
111915
indoline, 2,3-dihydroindole, 1-azaindan, dihydroindole, 1h-indole, 2,3-dihydro, azaindane, benzopyrrolidine, aza-indane, indoline, 19, pubchem7502
LPAGFVYQRIESJQ-UHFFFAOYSA-N
2,3-dihydro-1H-indole
10328
119.17
Indoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'einecsNumber' : 207-816-8

missing translation for 'rtecsNumber' : NL6906300

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only