missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Indomethacin Product Code.: 15462202

Thermo Scientific Chemicals Indomethacin

Product Code. 15462202
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15462202

Brand: Thermo Scientific Chemicals 458030050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indomethacin, CAS # 53-86-1, is an indole-based nonsteroidal anti-inflammatory (NSAID).

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Indomethacin is a synthetic NSAID of the indole family that inhibits cyclooxygenase and reduces prostaglandin and pyrogen release

• Indomethacin may suppress the multi-drug resistant transporter and thereby increase the efficacy of antineoplastics

• Indomethacin activates phosphatases that inhibit signaling pathways associated with cancer cell migration and proliferation

Applications

• Indomethacin may be used for the laboratory study of cancer cell proliferation, tumorigenesis and drug interactions

• Indomethacin may be used for laboratory study of the effect of inhibiting the multi-drug resistance transporter, or for downregulation of survival signaling in cells via survivin

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-86-1
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
InChI Key CGIGDMFJXJATDK-UHFFFAOYSA-N
Synonym indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
PubChem CID 3715
ChEBI CHEBI:49662
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O

Specifications

Melting Point 155.0°C
Loss on Drying 0.5% max. (100°C, 2 h, 5 mm Hg)
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 357.79
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Indomethacin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462819 Certificate TypeCertificate of Analysis Date28/05/2024 Catalog Number458030250,458031000,458030050
Lot NumberA0451821 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number458030250,458031000,458030050
Lot NumberA0407429 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number458030250,458031000,458030050
Lot NumberA0392380 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number458030250,458031000,458030050
Lot NumberA0414008 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number458030250,458031000,458030050
compliance-icons compliance-icons
Product Identifier
  • Indomethacin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H300-Fatal if swallowed.
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Indomethacin >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.