missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indoxyl acetate, 97%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 128.0°C to 131.0°C |
Color | Colorless to Lilac |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Quantity | 25 g |
Beilstein | 21, 71 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ethanol,chloroform |
Formula Weight | 175.19 |
Percent Purity | 97% |
Show More |
Safety and Handling
Product Identifier
- Indoxyl acetate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'