missing translation for 'onlineSavingsMsg'
Learn More

Infectious Mononucleosis Test using Latex Agglutination

Carry out specific detection of infectious mononucleosis heterophile antibody by latex agglutination.

Brand:  Thermo Scientific Oxoid DR0780M

Product Code. 11913132

  • 338.00 € / 100 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 8°C
Mononucleosis Antibodies
100 Tests
Infectious Mononucleosis Test
Test latex, positive control, negative control, disposable black slides, paddle pastettes, instruction for use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6186293 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberDR0780M
Lot Number6162843 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberDR0780M
Lot Number6136976 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberDR0780M
Lot Number6110939 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberDR0780M
Lot Number3801503 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog NumberDR0780M
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Infectious Mononucleosis Test using Latex Agglutination >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.