Learn More
Ingenol-3,4,5,20-diacetonide, MedChemExpress
MedChemExpress Ingenol-3,4,5,20-diacetonide is a natural compound.
Brand:  Medchem Express HY-N0871
Encompass_Preferred
 View more versions of this product
Product Code. 16471558
  • 905.85 € / 5mg
Explore more special offers
Description
Description
Ingenol-3,4,5,20-diacetonide is a natural compound.
Specifications
Specifications
Ingenol-3,4,5,20-diacetonide
White
C26H36O5
Ingenol 3,4:5,20-bisacetonide
428.56
98.19%
Solid
77573-44-5
Research
5 mg
O=C1[C@@]2(C=C(C)C3OC(C)(C)O4)C34C(OC(C)(C)OC5)C5=C[C@@H]1[C@@H](C6(C)C)[C@@H]6CC2C
428.56
Research